Assay Details
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019149.3 | NP_062022.2 | 5 - 6 | 1367 | 58 | |
XM_006254534.2 | XP_006254596.1 | 5 - 6 | 1389 | 58 | ||
XM_006254535.2 | XP_006254597.1 | 5 - 6 | 1340 | 58 | ||
GenBank mRNA | AB205483.1 | - | 4 - 5 | 2137 | 58 | |
BC062231.1 | - | 5 - 6 | 1390 | 58 | ||
BC168723.1 | - | 5 - 6 | 1339 | 58 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
