Assay Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022209.3 | NP_071545.2 | 3 - 4 | 537 | 75 | |
NR_073588.1 | 6 - 7 | 746 | 75 | |||
GenBank mRNA | AY251277.1 | - | 3 - 4 | 489 | 75 | |
BC078834.1 | - | 6 - 7 | 746 | 75 | ||
D14421.1 | - | 3 - 4 | 312 | 75 | ||
D38260.1 | - | 3 - 4 | 332 | 75 | ||
FM080850.1 | - | 3 - 4 | 537 | 75 | ||
FQ212581.1 | - | 1 - 2 | 57 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
