Assay Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022209.3 | NP_071545.2 | 5 - 6 | 831 | 91 | |
NR_073588.1 | 8 - 9 | 1040 | 91 | |||
GenBank mRNA | BC078834.1 | - | 8 - 9 | 1040 | 91 | |
D38260.1 | - | 5 - 6 | 626 | 91 | ||
FQ212581.1 | - | 3 - 4 | 351 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
