Assay Details
Gene Symbol: Hars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014012.2 | NP_001014034.2 | 6 - 7 | 758 | 112 | |
XM_006254575.2 | XP_006254637.1 | 5 - 6 | 668 | 112 | ||
GenBank mRNA | CV075096.1 | - | 3 - 4 | 361 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
