Assay Details
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012163.1 | NP_001012163.1 | 1 - 2 | 192 | 60 | |
XM_006254597.2 | XP_006254659.1 | 1 - 2 | 367 | 60 | ||
GenBank mRNA | BC087108.1 | - | 1 - 2 | 192 | 60 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087108.1, NM_001012163.1, XM_006254597.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
