Assay Details
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014147.1 | NP_001014169.2 | 1 - 2 | 166 | 79 | |
GenBank mRNA | BC079332.1 | - | 1 - 2 | 166 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079332.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
