Assay Details
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014147.1 | NP_001014169.2 | 3 - 4 | 330 | 78 | |
GenBank mRNA | BC079332.1 | - | 3 - 4 | 330 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
