Assay Details
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014147.1 | NP_001014169.2 | 6 - 7 | 734 | 81 | |
GenBank mRNA | BC079332.1 | - | 6 - 7 | 734 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P851797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176875 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P851797 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P851797 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P851797 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11192 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P165295 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P575385 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P851797 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29841 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103755 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36448 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26951 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10935034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803535 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803534 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10935033 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10803531 | - | - |
Rat Genome 230 2.0 Array | 1388532_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383270_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383270_at | - | - |
Rat Genome 230 2.0 Array | 1388532_at | - | - |
Affymetrix HT Rat Focus Array | 1388532_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383270_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388532_at | - | - |
Affymetrix HT Rat Focus Array | 1383270_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800008098.rn.1 | TC1800001136.rn.1 | RTA TC |
Rat Expression Set 230 | 1388532_at | - | - |
Rat Expression Set 230 | 1388532_PM_at | - | - |
Rat Expression Set 230 | 1383270_PM_at | - | - |
Rat Expression Set 230 | 1383270_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803531 | - | - |
Rat Genome U34 Set | rc_AI012108_at | - | - |
Rat Genome U34 Set | rc_AA957378_at | - | - |
Rat Genome U34 Set | rc_AI230038_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21486675 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349756 | - | - |
