Assay Details
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024741.1 | NP_001019912.1 | 10 - 11 | 1031 | 96 | |
XM_006248044.2 | XP_006248106.1 | 10 - 11 | 1067 | 96 | ||
GenBank mRNA | BC097460.1 | - | 10 - 11 | 1031 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
