Assay Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106791.2 | NP_001100261.2 | 7 - 8 | 1005 | 65 | |
XM_006242193.2 | XP_006242255.1 | 7 - 8 | 1073 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
