Assay Details
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033352.1 | NP_203503.1 | 1 - 2 | 1140 | 83 | |
GenBank mRNA | AF131294.1 | - | 1 - 2 | 1140 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
