Assay Details
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017177.1 | NP_058873.1 | 2 - 3 | 608 | 64 | |
XM_006242181.2 | XP_006242243.2 | 2 - 3 | 1260 | 64 | ||
XM_008765677.1 | XP_008763899.1 | 2 - 3 | 1271 | 64 | ||
GenBank mRNA | AB006607.1 | - | 2 - 3 | 608 | 64 | |
BC060515.1 | - | 2 - 3 | 584 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
