Assay Details
Gene Symbol: Creld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037208.1 | NP_001032285.1 | 6 - 7 | 723 | 54 | |
XM_006242216.2 | XP_006242278.1 | 5 - 6 | 578 | 54 | ||
GenBank mRNA | BC098722.1 | - | 6 - 7 | 723 | 54 | |
FQ229732.1 | - | 2 - 3 | 291 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
