Assay Details
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021746.1 | NP_068514.1 | 3 - 4 | 499 | 63 | |
XM_008765781.1 | XP_008764003.1 | 3 - 4 | 228 | 63 | ||
GenBank mRNA | X96488.1 | - | 3 - 4 | 499 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
