Assay Details
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145771.2 | NP_665714.2 | 3 - 4 | 182 | 90 | |
XM_008765674.1 | XP_008763896.1 | 3 - 4 | 366 | 90 | ||
GenBank mRNA | AF197128.1 | - | 3 - 4 | 176 | 90 | |
BC078840.1 | - | 3 - 4 | 182 | 90 | ||
FQ219187.1 | - | 2 - 3 | 171 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
