Assay Details
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145771.2 | NP_665714.2 | 7 - 8 | 587 | 88 | |
XM_008765674.1 | XP_008763896.1 | 7 - 8 | 771 | 88 | ||
GenBank mRNA | AF197128.1 | - | 7 - 8 | 581 | 88 | |
AF230096.1 | - | 6 - 7 | 589 | 88 | ||
AY738259.1 | - | 6 - 7 | 581 | 88 | ||
BC078840.1 | - | 7 - 8 | 587 | 88 | ||
FQ219187.1 | - | 6 - 7 | 576 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
