Assay Details
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079939.1 | NP_001073408.1 | 10 - 11 | 1579 | 64 | |
GenBank mRNA | BC129102.1 | - | 10 - 11 | 1579 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
