Assay Details
Gene Symbol: Hdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053447.1 | NP_445899.1 | 7 - 8 | 936 | 81 | |
GenBank mRNA | BC161939.1 | - | 7 - 8 | 911 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
