Assay Details
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108536.1 | NP_001102006.1 | 4 - 5 | 534 | 143 | |
XM_006256528.2 | XP_006256590.1 | 5 - 6 | 452 | 143 | ||
XM_006256529.1 | XP_006256591.1 | 4 - 5 | 322 | 143 | ||
GenBank mRNA | BC158720.1 | - | 5 - 6 | 436 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
