Assay Details
Gene Symbol: Frk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024368.1 | NP_077344.1 | 3 - 4 | 1073 | 114 | |
XM_006256517.2 | XP_006256579.1 | 3 - 4 | 1224 | 114 | ||
XM_008773006.1 | XP_008771228.1 | 3 - 4 | 1224 | 114 | ||
GenBank mRNA | U09583.1 | - | 3 - 4 | 1073 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
