Assay Details
Gene Symbol: Nmrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750277.3 | XP_003750325.2 | 4 - 5 | 606 | 65 | |
XM_008765147.1 | XP_008763369.1 | 4 - 5 | 606 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
