Assay Details
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109882.1 | NP_001103352.1 | 1 - 2 | 158 | 67 | |
GenBank mRNA | AY120888.1 | - | 1 - 2 | 77 | 67 | |
CV119278.1 | - | 1 - 2 | 158 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
