Assay Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021762.1 | NP_068530.1 | 2 - 3 | 271 | 108 | |
XM_008769496.1 | XP_008767718.1 | 2 - 3 | 290 | 108 | ||
GenBank mRNA | AF262356.1 | - | 2 - 3 | 271 | 108 | |
AY325258.1 | - | 346 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
