Assay Details
Gene Symbol: Kdm5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773146.1 | XP_008771368.1 | 27 - 28 | 4833 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071401 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071401 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071401 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071401 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P363672 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10933198 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933191 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933200 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933188 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933189 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933199 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933187 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933190 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933197 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933192 | - | - |
Rat Genome 230 2.0 Array | 1390586_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393093_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393093_at | - | - |
Rat Genome 230 2.0 Array | 1393199_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393199_at | - | - |
Rat Genome 230 2.0 Array | 1390586_at | - | - |
Affymetrix HT Rat Focus Array | 1393199_at | - | - |
Affymetrix HT Rat Focus Array | 1390586_at | - | - |
Affymetrix HT Rat Focus Array | 1390586_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393199_PM_at | - | - |
Rat Expression Set 230 | 1390586_at | - | - |
Rat Expression Set 230 | 1393093_PM_at | - | - |
Rat Expression Set 230 | 1393093_at | - | - |
Rat Expression Set 230 | 1390586_PM_at | - | - |
Rat Expression Set 230 | 1393199_PM_at | - | - |
Rat Expression Set 230 | 1393199_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933186 | - | - |
Rat Genome U34 Set | rc_AA926075_at | - | - |
Rat Genome U34 Set | rc_AA955145_at | - | - |
Rat Genome U34 Set | rc_AI177489_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21240079 | - | - |
ABI Rat Genome Survey Microarray | 22378072 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368975 | - | - |
RatRef-12 Expression BeadChip | ILMN_70069 | - | - |
