Assay Details
Gene Symbol: Kdm5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773146.1 | XP_008771368.1 | 21 - 22 | 3521 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
