Assay Details
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107452.1 | NP_001100922.1 | 1 - 2 | 223 | 66 | |
XM_006227784.2 | XP_006227846.1 | 1 - 2 | 212 | 66 | ||
GenBank mRNA | BC168744.1 | - | 1 - 2 | 180 | 66 | |
FQ213638.1 | - | 1 - 2 | 144 | 66 | ||
FQ219953.1 | - | 1 - 2 | 144 | 66 | ||
FQ219997.1 | - | 1 - 2 | 144 | 66 | ||
FQ231349.1 | - | 1 - 2 | 144 | 66 | ||
FQ231760.1 | - | 1 - 2 | 144 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
