Assay Details
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107452.1 | NP_001100922.1 | 5 - 6 | 601 | 85 | |
XM_006227784.2 | XP_006227846.1 | 5 - 6 | 584 | 85 | ||
GenBank mRNA | BC168744.1 | - | 5 - 6 | 552 | 85 | |
FQ213638.1 | - | 5 - 6 | 522 | 85 | ||
FQ219953.1 | - | 5 - 6 | 516 | 85 | ||
FQ219997.1 | - | 5 - 6 | 522 | 85 | ||
FQ231349.1 | - | 5 - 6 | 522 | 85 | ||
FQ231760.1 | - | 5 - 6 | 522 | 85 | ||
FQ234686.1 | - | 5 - 6 | 522 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
