Assay Details
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019289.2 | NP_062162.1 | 1 - 2 | 72 | 73 | |
GenBank mRNA | BC062027.1 | - | 1 - 2 | 72 | 73 | |
FQ228791.1 | - | 1 - 2 | 43 | 73 | ||
FQ228917.1 | - | 1 - 2 | 88 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC062027.1, NM_019289.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
