Assay Details
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017065.1 | NP_058761.1 | 3 - 4 | 532 | 95 | |
GenBank mRNA | AY742860.1 | - | 862 | 95 | ||
X15468.1 | - | 3 - 4 | 532 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
