Assay Details
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104613.1 | NP_001098083.1 | 6 - 7 | 692 | 64 | |
GenBank mRNA | AB006815.1 | - | 5 - 6 | 656 | 64 | |
AB006816.1 | - | 6 - 7 | 692 | 64 | ||
AB006817.1 | - | 5 - 6 | 656 | 64 | ||
AB006818.1 | - | 6 - 7 | 692 | 64 | ||
BC166548.1 | - | 287 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
