Assay Details
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104613.1 | NP_001098083.1 | 9 - 10 | 1000 | 78 | |
GenBank mRNA | AB006815.1 | - | 8 - 9 | 964 | 78 | |
AB006816.1 | - | 9 - 10 | 1000 | 78 | ||
AB006817.1 | - | 7 - 8 | 844 | 78 | ||
AB006818.1 | - | 8 - 9 | 880 | 78 | ||
BC166548.1 | - | 475 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
