Assay Details
Gene Symbol: Tm9sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231413.1 | XP_006231475.1 | 8 - 9 | 1453 | 65 | |
GenBank mRNA | FQ209899.1 | - | 5 - 6 | 568 | 65 | |
FQ229341.1 | - | 4 - 5 | 477 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
