Assay Details
Gene Symbol: Gucy2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139042.2 | NP_620611.2 | 14 - 15 | 2329 | 120 | |
GenBank mRNA | AF024622.1 | - | 14 - 15 | 2329 | 120 | |
U33847.1 | - | 422 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
