Assay Details
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057206.1 | NP_476554.1 | 3 - 4 | 444 | 89 | |
GenBank mRNA | L09216.1 | - | 3 - 4 | 444 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
