Assay Details
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011942.1 | NP_001011942.1 | 1 - 2 | 1731 | 132 | |
XM_006231471.2 | XP_006231533.1 | 1 - 2 | 1824 | 132 | ||
XM_008760413.1 | XP_008758635.1 | 1 - 2 | 1825 | 132 | ||
GenBank mRNA | BC085930.1 | - | 1 - 2 | 1731 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
