Assay Details
Gene Symbol: Gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012071.1 | NP_001012071.1 | 4 - 5 | 647 | 96 | |
XM_006231590.2 | XP_006231652.1 | 4 - 5 | 609 | 96 | ||
XM_006231591.2 | XP_006231653.1 | 4 - 5 | 581 | 96 | ||
XM_006231592.1 | XP_006231654.1 | 4 - 5 | 1339 | 96 | ||
GenBank mRNA | BC079295.1 | - | 4 - 5 | 647 | 96 | |
FJ179406.1 | - | 2 - 3 | 149 | 96 |
Gene Symbol: LOC103690044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760499.1 | XP_008758721.1 | 2 - 3 | 1316 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
