Assay Details
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014168.1 | NP_001014190.1 | 3 - 4 | 454 | 87 | |
XM_006231545.2 | XP_006231607.1 | 2 - 3 | 360 | 87 | ||
GenBank mRNA | BC085841.1 | - | 3 - 4 | 454 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
