Assay Details
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025400.3 | NP_001020571.1 | 3 - 4 | 294 | 68 | |
NM_001035234.2 | NP_001030311.1 | 3 - 4 | 456 | 68 | ||
XM_006231608.2 | XP_006231670.1 | 3 - 4 | 733 | 68 | ||
GenBank mRNA | BC097986.1 | - | 3 - 4 | 294 | 68 | |
EV778425.1 | - | 3 - 4 | 456 | 68 |
Gene Symbol: LOC100363406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768448.1 | XP_008766670.1 | 2 | 419 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
