Assay Details
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025400.3 | NP_001020571.1 | 4 - 5 | 447 | 75 | |
NM_001035234.2 | NP_001030311.1 | 4 - 5 | 609 | 75 | ||
NR_051995.1 | 4 - 5 | 626 | 75 | |||
XM_006231608.2 | XP_006231670.1 | 4 - 5 | 886 | 75 | ||
GenBank mRNA | BC097986.1 | - | 4 - 5 | 447 | 75 | |
BC098697.1 | - | 4 - 5 | 588 | 75 | ||
EV778425.1 | - | 4 - 5 | 609 | 75 | ||
FQ220088.1 | - | 3 - 4 | 390 | 75 |
Gene Symbol: LOC100363406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768448.1 | XP_008766670.1 | 2 | 572 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
