Assay Details
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107611.2 | NP_001101081.2 | 12 - 13 | 3488 | 76 | |
GenBank mRNA | EU562301.1 | - | 12 - 13 | 3488 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
