Assay Details
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107611.2 | NP_001101081.2 | 13 - 14 | 3612 | 151 | |
GenBank mRNA | EU562301.1 | - | 13 - 14 | 3612 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P473812 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P126265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81661 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126265 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P473812 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126265 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P473812 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P473812 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P126265 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P473812 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46997 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10716124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716130 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716128 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716135 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10716118 | - | - |
Rat Genome 230 2.0 Array | 1378337_at | - | - |
Rat Genome 230 2.0 Array | 1391810_at | - | - |
Rat Genome 230 2.0 Array | 1391810_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378337_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378337_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391810_at | - | - |
Affymetrix HT Rat Focus Array | 1378337_at | - | - |
Affymetrix HT Rat Focus Array | 1391810_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100031254.rn.1 | TC0100003829.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100031257.rn.1 | TC0100003829.rn.1 | RTA TC |
Rat Expression Set 230 | 1391810_PM_at | - | - |
Rat Expression Set 230 | 1378337_at | - | - |
Rat Expression Set 230 | 1391810_at | - | - |
Rat Expression Set 230 | 1378337_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716118 | - | - |
Rat Genome U34 Set | rc_AI175541_at | - | - |
Rat Genome U34 Set | rc_AI071851_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21809119 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356385 | - | - |
