Assay Details
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108527.1 | NP_001101997.1 | 3 - 4 | 281 | 71 | |
XM_006231684.2 | XP_006231746.1 | 2 - 3 | 2342 | 71 | ||
XM_006231685.2 | XP_006231747.1 | 2 - 3 | 1178 | 71 | ||
GenBank mRNA | FQ215895.1 | - | 3 - 4 | 270 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
