Assay Details
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108527.1 | NP_001101997.1 | 5 - 6 | 361 | 133 | |
XM_006231684.2 | XP_006231746.1 | 4 - 5 | 2422 | 133 | ||
XM_006231685.2 | XP_006231747.1 | 4 - 5 | 1258 | 133 | ||
XM_008760590.1 | XP_008758812.1 | 3 - 4 | 2156 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
