Assay Details
Gene Symbol: Actr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106364.1 | NP_001099834.1 | 10 - 11 | 1089 | 89 | |
XM_006231470.2 | XP_006231532.1 | 10 - 11 | 1261 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1037131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71039 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1037131 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1037131 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1037131 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1037131 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 419 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51133 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10730579 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730580 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730583 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730577 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730576 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730581 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730585 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730582 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730584 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730578 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10730575 | - | - |
Rat Genome 230 2.0 Array | 1371741_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371741_at | - | - |
Rat Genome 230 2.0 Array | 1394438_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394438_at | - | - |
Affymetrix HT Rat Focus Array | 1371741_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371741_at | - | - |
Affymetrix HT Rat Focus Array | 1394438_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394438_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100063137.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063139.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063143.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063135.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063141.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063136.rn.1 | TC0100007818.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100063138.rn.1 | TC0100007818.rn.1 | RTA TC |
Rat Expression Set 230 | 1371741_PM_at | - | - |
Rat Expression Set 230 | 1371741_at | - | - |
Rat Expression Set 230 | 1394438_at | - | - |
Rat Expression Set 230 | 1394438_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10730575 | - | - |
Rat Genome U34 Set | rc_AA926303_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21391194 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357207 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040250 | - | - |
