Assay Details
Gene Symbol: Hhat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769894.1 | XP_008768116.1 | 7 - 8 | 1765 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
