Assay Details
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053796.1 | NP_446248.1 | 4 - 5 | 459 | 119 | |
GenBank mRNA | AF276998.1 | - | 4 - 5 | 459 | 119 | |
BC065309.1 | - | 4 - 5 | 441 | 119 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
