Assay Details
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053796.1 | NP_446248.1 | 5 - 6 | 664 | 92 | |
GenBank mRNA | AF241261.1 | - | 1 - 2 | 211 | 92 | |
AF276998.1 | - | 5 - 6 | 664 | 92 | ||
BC065309.1 | - | 5 - 6 | 646 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
