Assay Details
Gene Symbol: Itpkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019312.2 | NP_062185.2 | 5 - 6 | 2906 | 117 | |
GenBank mRNA | AJ242781.1 | - | 2455 | 117 | ||
X74227.1 | - | 2229 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
