Assay Details
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011907.1 | NP_001011907.1 | 11 - 12 | 1302 | 93 | |
XM_003751277.2 | XP_003751325.1 | 11 - 12 | 1315 | 93 | ||
GenBank mRNA | BC082067.1 | - | 11 - 12 | 1302 | 93 | |
FQ215922.1 | - | 11 - 12 | 1306 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
