Assay Details
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011907.1 | NP_001011907.1 | 9 - 10 | 1125 | 93 | |
XM_003751277.2 | XP_003751325.1 | 9 - 10 | 1138 | 93 | ||
GenBank mRNA | BC082067.1 | - | 9 - 10 | 1125 | 93 | |
FQ215922.1 | - | 9 - 10 | 1129 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
